missing translation for 'onlineSavingsMsg'
Learn More

(Phenylhydrazono)malononitrile 96.0+%, TCI America™

Catalog No. p-7135531
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23465G 5 g
P234625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23465G Supplier TCI America Supplier No. P23465G
Only null left

Chemical Identifiers

CAS 306-18-3
Molecular Formula C9H6N4
Molecular Weight (g/mol) 170.175
MDL Number MFCD00195983
InChI Key MENUYOGJCXAFFU-UHFFFAOYSA-N
PubChem CID 67541
IUPAC Name 2-(phenylhydrazinylidene)propanedinitrile
SMILES C1=CC=C(C=C1)NN=C(C#N)C#N

Specifications

Melting Point 133°C
Color Yellow
Quantity 5 g
UN Number 3439
Formula Weight 170.18
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (Phenylhydrazono)malononitrile