accessibility menu, dialog, popup

UserName

CAS RN 306-18-3

NNNHN

CAS RN 306-18-3

IUPAC Name: 1-cyano-N-phenylmethanecarbohydrazonoyl cyanide
Synonyms: carbonylcyanide,phenylhydrazone carbonyl cyanide,phenylhydrazone
Molecular Weight (g/mol): 170.18
Molecular Formula: C9H6N4
InChi Key: MENUYOGJCXAFFU-UHFFFAOYSA-N
SMILES: N#CC(=NNC1=CC=CC=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results