missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Phenylhydrazono)malononitrile 96.0+%, TCI America™
Specifications
Specifications
| Melting Point | 133°C |
| Color | Yellow |
| Quantity | 5 g |
| UN Number | 3439 |
| Formula Weight | 170.18 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (Phenylhydrazono)malononitrile |
Safety and Handling
| TSCA | No |