missing translation for 'onlineSavingsMsg'
Learn More

Phenylhydrazinium Chloride, Reag. Ph Eur, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. p-4939083
Change view
Click to view available options
Quantity:
100 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1072530100 100 g Glass bottle
M1072530250 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1072530100 Supplier Technidata Supplier No. 1.07253.0100
Only null left
For analysis

Chemical Identifiers

CAS 59-88-1
Molecular Formula C6H9ClN2
Molecular Weight (g/mol) 144.60
MDL Number MFCD00012924
InChI Key JOVOSQBPPZZESK-UHFFFAOYSA-N
Synonym phenylhydrazine hydrochloride, phenylhydrazinium chloride, phenylhydrazine-hcl, phenylhydrazine monohydrochloride, phenylhydrazine hcl, hydrazine, phenyl-, monohydrochloride, unii-h7qfk49svd, phenylhydrazin hydrochlorid, phenylhydrazine.hcl, ccris 5707
PubChem CID 60962
IUPAC Name hydrogen phenylhydrazine chloride
SMILES [H+].[Cl-].NNC1=CC=CC=C1

Specifications

Packaging Glass bottle
Quantity 100 g
UN Number UN 2811
Formula Weight 144.6
Grade Reagent Ph. Eur.
Physical Form Solid
Chemical Name or Material Phenylhydrazinium Chloride
Hazard Category H350: May cause cancer.
H341: Suspected of causing genetic defects.
H331: Toxic if inhaled.
H311: Toxic in contact with skin.
H301: Toxic if swallowed.
H372: Causes damage to organs through prolonged or repeated exposure.
H319: Causes serious eye irritation.
H315: Causes skin irritation.
H317: May cause an allergic skin reaction.
H400: Very toxic to aquatic life.
DOTInformation DOT Class: 6.1, Packaging Group: II
EINECSNumber 200-444-7