missing translation for 'onlineSavingsMsg'
Learn More

Phenylhydrazine Hydrochloride, 99+%

Catalog No. p-182873
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC151551000 100 g Plastic bottle
AC151555000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC151551000 Supplier Thermo Scientific Chemicals Supplier No. 15155 1000

CAS: 59-88-1 | C6H9ClN2 | 144.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59-88-1
Molecular Formula C6H9ClN2
Molecular Weight (g/mol) 144.60
MDL Number MFCD00012924
InChI Key JOVOSQBPPZZESK-UHFFFAOYSA-N
Synonym phenylhydrazine hydrochloride, phenylhydrazinium chloride, phenylhydrazine-hcl, phenylhydrazine monohydrochloride, phenylhydrazine hcl, hydrazine, phenyl-, monohydrochloride, unii-h7qfk49svd, phenylhydrazin hydrochlorid, phenylhydrazine.hcl, ccris 5707
PubChem CID 60962
IUPAC Name phenylhydrazine;hydrochloride
SMILES [H+].[Cl-].NNC1=CC=CC=C1

Specifications

Melting Point 250.0°C to 254.0°C
Color Beige-Pink or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Plastic bottle
Linear Formula C6H5NHNH2·HCl
Quantity 100 g
Beilstein 15, 67
Fieser 07,284
Merck Index 15, 7404
Solubility Information (5% in H2O) Clear colorless to light yellow solution
Formula Weight 144.61
Percent Purity 99+%
Physical Form Fine Crystalline Powder
Chemical Name or Material Phenylhydrazine hydrochloride, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if inhaled. Toxic if swallowed. Toxic in contact with skin. Suspected of causing genetic defects. May cause cancer. May cause an allergic skin reaction. Causes skin irritation. Causes serious eye irritation. Causes damage to organs through prolonged or repeated exposure. Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF exposed or concerned: Get medical advice/attention. Avoid release to the environment.
EINECSNumber 200-444-7
RTECSNumber MV9000000
TSCA TSCA

RUO – Research Use Only