missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylbis(2,4,6-trimethylbenzoyl)phosphine Oxide 96.0+%, TCI America™
Supplier: TCI America P231225G
Specifications
| Phenylbis(2,4,6-trimethylbenzoyl)phosphine Oxide | |
| 131°C | |
| C26H27O3P | |
| 25 g | |
| GUCYFKSBFREPBC-UHFFFAOYSA-N | |
| [phenyl-(2,4,6-trimethylbenzoyl)phosphoryl]-(2,4,6-trimethylphenyl)methanone | |
| 164512 | |
| ≥96.0% (GC) |
| 162881-26-7 | |
| Yellow | |
| MFCD01863675 | |
| BAPO | |
| CC1=CC(=C(C(=C1)C)C(=O)P(=O)(C2=CC=CC=C2)C(=O)C3=C(C=C(C=C3C)C)C)C | |
| 418.473 | |
| 418.47 | |
| Crystalline Powder |
Chemical Identifiers
| 162881-26-7 | |
| 418.473 | |
| GUCYFKSBFREPBC-UHFFFAOYSA-N | |
| 164512 | |
| CC1=CC(=C(C(=C1)C)C(=O)P(=O)(C2=CC=CC=C2)C(=O)C3=C(C=C(C=C3C)C)C)C |
| C26H27O3P | |
| MFCD01863675 | |
| BAPO | |
| [phenyl-(2,4,6-trimethylbenzoyl)phosphoryl]-(2,4,6-trimethylphenyl)methanone |
Safety and Handling
EINECSNumber : (3)-4445
TSCA : Yes