CAS RN 162881-26-7
IUPAC Name:
[phenyl(2,4,6-trimethylbenzoyl)phosphoryl](2,4,6-trimethylphenyl)methanone
Synonyms:
[phenyl(2,4,6-trimethylbenzoyl)phosphoryl](2,4,6-trimethylphenyl)methanone
Molecular Weight (g/mol):
418.47
Molecular Formula:
C26H27O3P
InChi Key:
GUCYFKSBFREPBC-UHFFFAOYSA-N
SMILES:
CC1=CC(C)=C(C(=O)P(=O)(C(=O)C2=C(C)C=C(C)C=C2C)C2=CC=CC=C2)C(C)=C1
1
–
2
of
2
results
1
Phenylbis(2,4,6-trimethylbenzoyl)phosphine Oxide 96.0+%, TCI America™
CAS: 162881-26-7 Molecular Formula: C26H27O3P Molecular Weight (g/mol): 418.473 MDL Number: MFCD01863675 InChI Key: GUCYFKSBFREPBC-UHFFFAOYSA-N Synonym: BAPO PubChem CID: 164512 IUPAC Name: [phenyl-(2,4,6-trimethylbenzoyl)phosphoryl]-(2,4,6-trimethylphenyl)methanone SMILES: CC1=CC(=C(C(=C1)C)C(=O)P(=O)(C2=CC=CC=C2)C(=O)C3=C(C=C(C=C3C)C)C)C
2
Sigma Aldrich Phenylbis(2,4,6-trimethylbenzoyl)phosphine oxide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.