missing translation for 'onlineSavingsMsg'
Learn More

Phenylbis(2,4,6-trimethylbenzoyl)phosphine Oxide 96.0+%, TCI America™

Catalog No. P23125G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23125G 5 g
P231225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23125G Supplier TCI America Supplier No. P23125G

Chemical Identifiers

CAS 162881-26-7
Molecular Formula C26H27O3P
Molecular Weight (g/mol) 418.473
MDL Number MFCD01863675
InChI Key GUCYFKSBFREPBC-UHFFFAOYSA-N
Synonym BAPO
PubChem CID 164512
IUPAC Name [phenyl-(2,4,6-trimethylbenzoyl)phosphoryl]-(2,4,6-trimethylphenyl)methanone
SMILES CC1=CC(=C(C(=C1)C)C(=O)P(=O)(C2=CC=CC=C2)C(=O)C3=C(C=C(C=C3C)C)C)C

Specifications

Melting Point 131°C
Color Yellow
Quantity 5 g
Formula Weight 418.47
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenylbis(2,4,6-trimethylbenzoyl)phosphine Oxide
EINECSNumber (3)-4445
TSCA Yes