missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylazoresorcinol 80.0+%, TCI America™
Supplier: TCI America P015125G
Specifications
| Phenylazoresorcinol | |
| C18H14N2O4 | |
| 25 g | |
| C1=CC=C(C=C1)C2=C(C=CC=C2ON=NOC3=CC=CC(=C3)O)O | |
| 322.32 | |
| 214.22 | |
| Crystalline Powder |
| 67503-46-2 | |
| MFCD00059881 | |
| KVQRGKQWGHNYJE-FMQUCBEESA-N | |
| 3-[(E)-(3-hydroxyphenoxy)diazenyl]oxy-2-phenylphenol | |
| 77519172 | |
| ≥80.0% (T) |
Chemical Identifiers
| 67503-46-2 | |
| 322.32 | |
| KVQRGKQWGHNYJE-FMQUCBEESA-N | |
| 3-[(E)-(3-hydroxyphenoxy)diazenyl]oxy-2-phenylphenol |
| C18H14N2O4 | |
| MFCD00059881 | |
| 77519172 | |
| C1=CC=C(C=C1)C2=C(C=CC=C2ON=NOC3=CC=CC(=C3)O)O |
Safety and Handling
TSCA : No