missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylazoresorcinol 80.0+%, TCI America™
$59.65 - $203.17
Chemical Identifiers
| CAS | 67503-46-2 |
|---|---|
| Molecular Formula | C18H14N2O4 |
| Molecular Weight (g/mol) | 322.32 |
| MDL Number | MFCD00059881 |
| InChI Key | KVQRGKQWGHNYJE-FMQUCBEESA-N |
| PubChem CID | 77519172 |
| IUPAC Name | 3-[(E)-(3-hydroxyphenoxy)diazenyl]oxy-2-phenylphenol |
| SMILES | C1=CC=C(C=C1)C2=C(C=CC=C2ON=NOC3=CC=CC(=C3)O)O |
Chemical Identifiers
| 67503-46-2 | |
| 322.32 | |
| KVQRGKQWGHNYJE-FMQUCBEESA-N | |
| 3-[(E)-(3-hydroxyphenoxy)diazenyl]oxy-2-phenylphenol |
| C18H14N2O4 | |
| MFCD00059881 | |
| 77519172 | |
| C1=CC=C(C=C1)C2=C(C=CC=C2ON=NOC3=CC=CC(=C3)O)O |
Specifications
| 67503-46-2 | |
| MFCD00059881 | |
| KVQRGKQWGHNYJE-FMQUCBEESA-N | |
| 3-[(E)-(3-hydroxyphenoxy)diazenyl]oxy-2-phenylphenol | |
| 77519172 | |
| ≥80.0% (T) | |
| Phenylazoresorcinol |
| C18H14N2O4 | |
| 1 g | |
| C1=CC=C(C=C1)C2=C(C=CC=C2ON=NOC3=CC=CC(=C3)O)O | |
| 322.32 | |
| 214.22 | |
| Crystalline Powder |
Safety and Handling
TSCA : No