missing translation for 'onlineSavingsMsg'
Learn More

Phenylazoformic acid 2-phenylhydrazide compound with 1,5-Diphenylcarbohydrazide, ACS

Catalog No. AA3644209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3644209 10 g
AA3644218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3644209 Supplier Thermo Scientific Chemicals Supplier No. 03644209

CAS: 10329-15-4 | C26H26N8O2 | 482.55 g/mol

Phenylazoformic acid 2-phenylhydrazide compound with 1,5-Diphenylcarbohydrazide is used as an indicator in titrating iron and for the colorimetric determination of chromium and the detection of cadmium, mercury, magnesium, aldehydes, and emetine, a color developing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10329-15-4
Molecular Formula C26H26N8O2
Molecular Weight (g/mol) 482.55
MDL Number MFCD00013218
InChI Key WDJPBIRJSVMEKJ-UHFFFAOYSA-N
Synonym 1-anilino-3-phenyliminourea; 1,3-dianilinourea, 1,5-diphenylcarbazide; 3-phenylamino-1-phenylimino urea, n',2-diphenyldiazenecarbohydrazide compound with n,n'-diphenylcarbonohydrazide 1:1, a.c.s. reagent cpd w/1 5-diph-carbazide, diphenylcarbazone compound with diphenylcarbazide, 3e-1-anilino-3-phenylimino-urea; 1,3-dianilinourea, diphenylcarbazone compound with s-diphenyl carbazide, acs, e-1-phenylamino-3-phenylimino urea; 1,5-diphenylcarbazide, e-3-phenylamino-1-phenylimino urea; 1,5-diphenylcarbazide, phenylazoformic acid 2-phenylhydrazide compound 1,5diphenylcarbohydrazide
PubChem CID 221124
IUPAC Name 1-anilino-3-phenyliminourea;1,3-dianilinourea
SMILES O=C(NNC1=CC=CC=C1)NNC1=CC=CC=C1.O=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1

Specifications

Melting Point 154°C to 156°C
Color Orange
Odor Odorless
Quantity 10 g
Beilstein 6110063
Merck Index 14,3322
Formula Weight 482.55
Grade ACS
Physical Form Powder
Chemical Name or Material Phenylazoformic acid 2-phenylhydrazide compound, with 1,5-Diphenylcarbohydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only