accessibility menu, dialog, popup

UserName

ONHHNNHHNONHHNNN(E/Z)

CAS RN 10329-15-4

IUPAC Name: 1,3-bis(phenylamino)urea; 3-(phenylamino)-1-(phenylimino)urea
Synonyms: 1,5-diphenylcarbazide; 3-(phenylamino)-1-(phenylimino)urea
Molecular Weight (g/mol): 482.55
Molecular Formula: C26H26N8O2
InChi Key: WDJPBIRJSVMEKJ-UHFFFAOYSA-N
SMILES: O=C(NNC1=CC=CC=C1)NNC1=CC=CC=C1.O=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results