missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl Vinyl Sulfoxide 95.0+%, TCI America™
$246.05 - $866.79
Chemical Identifiers
| CAS | 20451-53-0 |
|---|---|
| Molecular Formula | C8H8OS |
| Molecular Weight (g/mol) | 152.21 |
| MDL Number | MFCD00002086 |
| InChI Key | MZMJHXFYLRTLQX-UHFFFAOYNA-N |
| PubChem CID | 88544 |
| IUPAC Name | (ethenesulfinyl)benzene |
| SMILES | C=CS(=O)C1=CC=CC=C1 |
Chemical Identifiers
| 20451-53-0 | |
| 152.21 | |
| MZMJHXFYLRTLQX-UHFFFAOYNA-N | |
| (ethenesulfinyl)benzene |
| C8H8OS | |
| MFCD00002086 | |
| 88544 | |
| C=CS(=O)C1=CC=CC=C1 |
Specifications
| 20451-53-0 | |
| 110°C | |
| MFCD00002086 | |
| MZMJHXFYLRTLQX-UHFFFAOYNA-N | |
| (ethenesulfinyl)benzene | |
| 88544 | |
| ≥95.0% (GC) | |
| Phenyl Vinyl Sulfoxide |
| Yellow | |
| C8H8OS | |
| 5 g | |
| C=CS(=O)C1=CC=CC=C1 | |
| 152.21 | |
| 152.21 | |
| Liquid |
Safety and Handling
TSCA : No