missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Vinyl Sulfoxide 95.0+%, TCI America™

Catalog No. P086325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P086325G 25 g
P08635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P086325G Supplier TCI America Supplier No. P086325G

Chemical Identifiers

CAS 20451-53-0
Molecular Formula C8H8OS
Molecular Weight (g/mol) 152.21
MDL Number MFCD00002086
InChI Key MZMJHXFYLRTLQX-UHFFFAOYNA-N
PubChem CID 88544
IUPAC Name (ethenesulfinyl)benzene
SMILES C=CS(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 110°C
Quantity 25 g
Formula Weight 152.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenyl Vinyl Sulfoxide