missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Vinyl Sulfide (stabilized with TBC) 98.0+%, TCI America™

Catalog No. P12875G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12875G 5 g
P128725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12875G Supplier TCI America Supplier No. P12875G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1822-73-7
Molecular Formula C8H8S
Molecular Weight (g/mol) 136.21
MDL Number MFCD00009646
InChI Key GMPDOIGGGXSAPL-UHFFFAOYSA-N
Synonym phenyl vinyl sulfide, phenylthioethene, phenyl vinyl sulphide, phenylthio ethylene, vinylthiobenzene, benzene, ethenylthio, vinyl phenyl sulfide, sulfide, phenyl vinyl, vinylsulfanyl benzene, phenyl vinyl thioether
PubChem CID 74572
IUPAC Name (ethenylsulfanyl)benzene
SMILES C=CSC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 78°C
Quantity 5 g
UN Number 1993
Formula Weight 136.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenyl Vinyl Sulfide (stabilized with TBC)