accessibility menu, dialog, popup

UserName

CH2S

CAS RN 1822-73-7

IUPAC Name: (ethenylsulfanyl)benzene
Synonyms: phenyl vinyl sulfide
Molecular Weight (g/mol): 136.21
Molecular Formula: C8H8S
InChi Key: GMPDOIGGGXSAPL-UHFFFAOYSA-N
SMILES: C=CSC1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)