missing translation for 'onlineSavingsMsg'
Learn More

Phenyl trans-beta-styryl sulfone, 96%

Catalog No. AAA1855606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1855606 5 g
AAA1855603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1855606 Supplier Thermo Scientific Chemicals Supplier No. A1855606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16212-06-9 | C14H12O2S | 244.308 g/mol

Phenyl trans-beta-styryl sulfone is used as a building block for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl trans-beta-styryl sulfone is used as a building block for proteomics research.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 16212-06-9
Molecular Formula C14H12O2S
Molecular Weight (g/mol) 244.308
MDL Number MFCD00159177
InChI Key DNMCCXFLTURVLK-VAWYXSNFSA-N
Synonym phenyl trans-styryl sulfone, phenyl styryl sulfone, sulfone, phenyl styryl, phenyl trans-styryl sulphone, phenyl 2-phenylvinyl sulfone, benzene, 2-phenylethenyl sulfonyl, e-2-benzenesulfonyl ethenyl benzene, 1e-2-phenylvinyl sulfonyl benzene, styrylsulfonylbenzene, e-2-phenylethenyl sulphonylbenzene
PubChem CID 736143
IUPAC Name [(E)-2-(benzenesulfonyl)ethenyl]benzene
SMILES C1=CC=C(C=C1)C=CS(=O)(=O)C2=CC=CC=C2

Specifications

Melting Point 74°C to 76°C
Quantity 5 g
Formula Weight 244.32
Percent Purity 98%
Chemical Name or Material Phenyl trans-beta-styryl sulfone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only