Learn More
Phenyl salicylate, 99%
CAS: 118-55-8 | C13H10O3 | 214.22 g/mol
Supplier: Thermo Scientific Chemicals B2068636
Description
Phenyl salicylate is used in the production of polymers, lacquers, adhesives, waxes and polishes. It acts as an antiseptic and mild analgesic. It is also used in the prevention of sunburn. It reacts with o-toluidine in 1,2,4-trichlorobenzene to give the corresponding amide o-Salicylotoluide. Further, it is involved in the study to check the effect on crystal size with cooling rate in the laboratory. In addition to this, it is used in veterinary medicine and also employed for coating pills, in which the medicine is intended for enteric release salol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenyl salicylate is used in the production of polymers, lacquers, adhesives, waxes and polishes. It acts as an antiseptic and mild analgesic. It is also used in the prevention of sunburn. It reacts with o-toluidine in 1,2,4-trichlorobenzene to give the corresponding amide o-Salicylotoluide. Further, it is involved in the study to check the effect on crystal size with cooling rate in the laboratory. In addition to this, it is used in veterinary medicine and also employed for coating pills, in which the medicine is intended for enteric release salol.
Solubility
Soluble in alcohol, ether, chloroform, turpentine, acetone and benzene. Sparingly soluble in chloroformbenzene. Insoluble in water.
Notes
Light sensitive. Incompatible with strong oxidants, strong acids and strong bases. Store in a cool place.
Specifications
| Phenyl salicylate | |
| 41°C to 45°C | |
| 160°C (320°F) | |
| MFCD00002213 | |
| 393969 | |
| phenyl salicylate, salol, phenol salicylate, salphenyl, musol, 2-phenoxycarbonylphenol, benzoic acid, 2-hydroxy-, phenyl ester, salicylic acid, phenyl ester, 2-hydroxybenzoic acid phenyl ester, phenyl-2-hydroxybenzoate | |
| ZQBAKBUEJOMQEX-UHFFFAOYSA-N | |
| phenyl 2-hydroxybenzoate | |
| 8361 | |
| 214.22 |
| 118-55-8 | |
| 172°C to 174°C (12 mmHg) | |
| C13H10O3 | |
| 500 g | |
| 14,7310 | |
| Soluble in alcohol,ether,chloroform,turpentine,acetone and benzene. Sparingly soluble in chloroformbenzene. Insoluble in water. | |
| C1=CC=C(C=C1)OC(=O)C2=CC=CC=C2O | |
| 214.22 | |
| CHEBI:34918 | |
| 99% |
Chemical Identifiers
| 118-55-8 | |
| 214.22 | |
| ZQBAKBUEJOMQEX-UHFFFAOYSA-N | |
| 8361 | |
| phenyl 2-hydroxybenzoate |
| C13H10O3 | |
| MFCD00002213 | |
| phenyl salicylate, salol, phenol salicylate, salphenyl, musol, 2-phenoxycarbonylphenol, benzoic acid, 2-hydroxy-, phenyl ester, salicylic acid, phenyl ester, 2-hydroxybenzoic acid phenyl ester, phenyl-2-hydroxybenzoate | |
| CHEBI:34918 | |
| C1=CC=C(C=C1)OC(=O)C2=CC=CC=C2O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 204-259-2
RTECSNumber : VO6125000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only