missing translation for 'onlineSavingsMsg'
Learn More

Phenyl salicylate, 99%

Catalog No. p-7025286
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2068622 100 g
AAB2068636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2068622 Supplier Thermo Scientific Chemicals Supplier No. B2068622
Only null left

CAS: 118-55-8 | C13H10O3 | 214.22 g/mol

Phenyl salicylate is used in the production of polymers, lacquers, adhesives, waxes and polishes. It acts as an antiseptic and mild analgesic. It is also used in the prevention of sunburn. It reacts with o-toluidine in 1,2,4-trichlorobenzene to give the corresponding amide o-Salicylotoluide. Further, it is involved in the study to check the effect on crystal size with cooling rate in the laboratory. In addition to this, it is used in veterinary medicine and also employed for coating pills, in which the medicine is intended for enteric release salol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl salicylate is used in the production of polymers, lacquers, adhesives, waxes and polishes. It acts as an antiseptic and mild analgesic. It is also used in the prevention of sunburn. It reacts with o-toluidine in 1,2,4-trichlorobenzene to give the corresponding amide o-Salicylotoluide. Further, it is involved in the study to check the effect on crystal size with cooling rate in the laboratory. In addition to this, it is used in veterinary medicine and also employed for coating pills, in which the medicine is intended for enteric release salol.

Solubility
Soluble in alcohol, ether, chloroform, turpentine, acetone and benzene. Sparingly soluble in chloroformbenzene. Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidants, strong acids and strong bases. Store in a cool place.

Chemical Identifiers

CAS 118-55-8
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00002213
InChI Key ZQBAKBUEJOMQEX-UHFFFAOYSA-N
Synonym phenyl salicylate, salol, phenol salicylate, salphenyl, musol, 2-phenoxycarbonylphenol, benzoic acid, 2-hydroxy-, phenyl ester, salicylic acid, phenyl ester, 2-hydroxybenzoic acid phenyl ester, phenyl-2-hydroxybenzoate
PubChem CID 8361
ChEBI CHEBI:34918
IUPAC Name phenyl 2-hydroxybenzoate
SMILES C1=CC=C(C=C1)OC(=O)C2=CC=CC=C2O

Specifications

Melting Point 41°C to 45°C
Boiling Point 172°C to 174°C (12 mmHg)
Flash Point 160°C (320°F)
Quantity 100 g
Beilstein 393969
Merck Index 14,7310
Solubility Information Soluble in alcohol,ether,chloroform,turpentine,acetone and benzene. Sparingly soluble in chloroformbenzene. Insoluble in water.
Formula Weight 214.22
Percent Purity 99%
Chemical Name or Material Phenyl salicylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-259-2
RTECSNumber VO6125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only