missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Propionate 97.0+%, TCI America™

Catalog No. p-7134649
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P050925G 25 g
P0509100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P050925G Supplier TCI America Supplier No. P050925G

Chemical Identifiers

CAS 637-27-4
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00053765
InChI Key DYUMLJSJISTVPV-UHFFFAOYSA-N
Synonym Propionic Acid Phenyl Ester
PubChem CID 12497
IUPAC Name phenyl propanoate
SMILES CCC(=O)OC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 211°C
Quantity 25 g
Formula Weight 150.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenyl Propionate
EINECSNumber (3)-0643
TSCA Yes