CAS RN 637-27-4
IUPAC Name:
phenyl propanoate
Synonyms:
propanoic acid, phenyl ester
Molecular Weight (g/mol):
150.18
Molecular Formula:
C9H10O2
InChi Key:
DYUMLJSJISTVPV-UHFFFAOYSA-N
SMILES:
CCC(=O)OC1=CC=CC=C1
1
–
1
of
1
results
1
Phenyl Propionate 97.0+%, TCI America™
CAS: 637-27-4 Molecular Formula: C9H10O2 Molecular Weight (g/mol): 150.177 MDL Number: MFCD00053765 InChI Key: DYUMLJSJISTVPV-UHFFFAOYSA-N Synonym: Propionic Acid Phenyl Ester PubChem CID: 12497 IUPAC Name: phenyl propanoate SMILES: CCC(=O)OC1=CC=CC=C1