missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl Methanesulfonate 98.0+%, TCI America™
$257.97 - $257.97
Chemical Identifiers
| CAS | 16156-59-5 |
|---|---|
| Molecular Formula | C7H8O3S |
| Molecular Weight (g/mol) | 172.20 |
| MDL Number | MFCD00095143 |
| InChI Key | WXVUCMFEGJUVTN-UHFFFAOYSA-N |
| Synonym | phenyl methylsulfonate, phenyl methyl sulfonate, acmc-1bs7l, phenylmethanesulfonate 4, phenyl methanesulfonate, methanesulfonic acid,phenyl ester, methanesulfonic acid phenyl ester, methanesulfonic acid, phenyl ester |
| PubChem CID | 316170 |
| IUPAC Name | phenyl methanesulfonate |
| SMILES | CS(=O)(=O)OC1=CC=CC=C1 |
Chemical Identifiers
| 16156-59-5 | |
| 172.20 | |
| WXVUCMFEGJUVTN-UHFFFAOYSA-N | |
| 316170 | |
| CS(=O)(=O)OC1=CC=CC=C1 |
| C7H8O3S | |
| MFCD00095143 | |
| phenyl methylsulfonate, phenyl methyl sulfonate, acmc-1bs7l, phenylmethanesulfonate 4, phenyl methanesulfonate, methanesulfonic acid,phenyl ester, methanesulfonic acid phenyl ester, methanesulfonic acid, phenyl ester | |
| phenyl methanesulfonate |
Specifications
| 16156-59-5 | |
| White-Yellow | |
| C7H8O3S | |
| 5 g | |
| WXVUCMFEGJUVTN-UHFFFAOYSA-N | |
| phenyl methanesulfonate | |
| 316170 | |
| ≥98.0% (GC) | |
| Phenyl Methanesulfonate |
| 61°C | |
| 279°C | |
| MFCD00095143 | |
| phenyl methylsulfonate, phenyl methyl sulfonate, acmc-1bs7l, phenylmethanesulfonate 4, phenyl methanesulfonate, methanesulfonic acid,phenyl ester, methanesulfonic acid phenyl ester, methanesulfonic acid, phenyl ester | |
| CS(=O)(=O)OC1=CC=CC=C1 | |
| 172.20 | |
| 172.20 | |
| Crystalline Powder |
Safety and Handling
TSCA : No