missing translation for 'onlineSavingsMsg'
Learn More

Phenyl methanesulfonate, 98%

CAS: 16156-59-5 | C7H8O3S | 172.20 g/mol

$37.60 - $103.27

Chemical Identifiers

CAS 16156-59-5
Molecular Formula C7H8O3S
Molecular Weight (g/mol) 172.20
MDL Number MFCD00095143
InChI Key WXVUCMFEGJUVTN-UHFFFAOYSA-N
Synonym phenyl methylsulfonate, phenyl methyl sulfonate, acmc-1bs7l, phenylmethanesulfonate 4, phenyl methanesulfonate, methanesulfonic acid,phenyl ester, methanesulfonic acid phenyl ester, methanesulfonic acid, phenyl ester
PubChem CID 316170
IUPAC Name phenyl methanesulfonate
SMILES CS(=O)(=O)OC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5599503
View Documents
Thermo Scientific Chemicals
H5599503
1 g
Each for $37.60
Only null left
 
AAH5599506
View Documents
Thermo Scientific Chemicals
H5599506
5 g
Each for $103.27
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

16156-59-5
172.20
WXVUCMFEGJUVTN-UHFFFAOYSA-N
316170
CS(=O)(=O)OC1=CC=CC=C1
C7H8O3S
MFCD00095143
phenyl methylsulfonate, phenyl methyl sulfonate, acmc-1bs7l, phenylmethanesulfonate 4, phenyl methanesulfonate, methanesulfonic acid,phenyl ester, methanesulfonic acid phenyl ester, methanesulfonic acid, phenyl ester
phenyl methanesulfonate

Specifications

16156-59-5
Brown
C7H8O3S
MFCD00095143
phenyl methylsulfonate, phenyl methyl sulfonate, acmc-1bs7l, phenylmethanesulfonate 4, phenyl methanesulfonate, methanesulfonic acid,phenyl ester, methanesulfonic acid phenyl ester, methanesulfonic acid, phenyl ester
CS(=O)(=O)OC1=CC=CC=C1
172.20
172.2
Solid
58°C to 61°C
279°C
CH3SO3C6H5
1 g
WXVUCMFEGJUVTN-UHFFFAOYSA-N
phenyl methanesulfonate
316170
98%
Phenyl methanesulfonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only