missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl Isothiocyanate 98.0+%, TCI America™
Supplier: TCI America I0191100G
Specifications
| Phenyl Isothiocyanate | |
| Yellow | |
| C7H5NS | |
| 100 g | |
| phenyl isothiocyanate, phenylisothiocyanate, thiocarbanil, phenyl mustard oil, benzene, isothiocyanato, pitc, isothiocyanic acid phenyl ester, benzene-1-isothiocyanate, phenylsenfoel, phenyl thioisocyanate | |
| S=C=NC1=CC=CC=C1 | |
| 135.18 | |
| CHEBI:85103 | |
| ≥98.0% (GC) |
| 103-72-0 | |
| 71°C | |
| MFCD00004798 | |
| 2922 | |
| QKFJKGMPGYROCL-UHFFFAOYSA-N | |
| isothiocyanatobenzene | |
| 7673 | |
| 135.18 | |
| Liquid |
Chemical Identifiers
| 103-72-0 | |
| 135.18 | |
| QKFJKGMPGYROCL-UHFFFAOYSA-N | |
| 7673 | |
| isothiocyanatobenzene |
| C7H5NS | |
| MFCD00004798 | |
| phenyl isothiocyanate, phenylisothiocyanate, thiocarbanil, phenyl mustard oil, benzene, isothiocyanato, pitc, isothiocyanic acid phenyl ester, benzene-1-isothiocyanate, phenylsenfoel, phenyl thioisocyanate | |
| CHEBI:85103 | |
| S=C=NC1=CC=CC=C1 |
Safety and Handling
EINECSNumber : (3)-3239
RTECSNumber : NX9275000
TSCA : Yes
Recommended Storage : Refrigerator