accessibility menu, dialog, popup

UserName

SCN

CAS RN 103-72-0

IUPAC Name: isothiocyanatobenzene
Synonyms: phenylisothiocyanate
Molecular Weight (g/mol): 135.18
Molecular Formula: C7H5NS
InChi Key: QKFJKGMPGYROCL-UHFFFAOYSA-N
SMILES: S=C=NC1=CC=CC=C1
Boiling Point 
  • (3)
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (1)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (6)
  • (3)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (7)