missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Isothiocyanate 98.0+%, TCI America™

Catalog No. I019125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I019125G 25 g
I0191100G 100 g
I0191500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I019125G Supplier TCI America Supplier No. I019125G

Chemical Identifiers

CAS 103-72-0
Molecular Formula C7H5NS
Molecular Weight (g/mol) 135.18
MDL Number MFCD00004798
InChI Key QKFJKGMPGYROCL-UHFFFAOYSA-N
Synonym phenyl isothiocyanate, phenylisothiocyanate, thiocarbanil, phenyl mustard oil, benzene, isothiocyanato, pitc, isothiocyanic acid phenyl ester, benzene-1-isothiocyanate, phenylsenfoel, phenyl thioisocyanate
PubChem CID 7673
ChEBI CHEBI:85103
IUPAC Name isothiocyanatobenzene
SMILES S=C=NC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 71°C
Quantity 25 g
UN Number 2922
Formula Weight 135.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenyl Isothiocyanate
EINECSNumber (3)-3239
RTECSNumber NX9275000
TSCA Yes
Recommended Storage Refrigerator