missing translation for 'onlineSavingsMsg'
Learn More

Phenyl isothiocyanate, 98%

Catalog No. AC160971000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160971000 100 g Glass bottle
AC160975000 500 g Glass bottle
AC160970025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC160971000 Supplier Thermo Scientific Chemicals Supplier No. 160971000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 103-72-0 | C7H5NS | 135.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103-72-0
Molecular Formula C7H5NS
Molecular Weight (g/mol) 135.18
MDL Number MFCD00004798
InChI Key QKFJKGMPGYROCL-UHFFFAOYSA-N
Synonym phenyl isothiocyanate, phenylisothiocyanate, thiocarbanil, phenyl mustard oil, benzene, isothiocyanato, pitc, isothiocyanic acid phenyl ester, benzene-1-isothiocyanate, phenylsenfoel, phenyl thioisocyanate
PubChem CID 7673
ChEBI CHEBI:85103
IUPAC Name isothiocyanatobenzene
SMILES S=C=NC1=CC=CC=C1

Specifications

Melting Point -21.0°C
Color Yellow
Density 1.1288g/mL
Boiling Point 221.0°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6490 to 1.6510
Linear Formula C6H5NCS
Quantity 100 g
Beilstein 12,453
Fieser 01,844; 02,323
Merck Index 15,7408
Specific Gravity 1.1288
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and diethyl ether
Viscosity 1.3 mPa.s (20°C)
Formula Weight 135.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Phenyl isothiocyanate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 203-138-1

RUO – Research Use Only