missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl Chlorothionoformate 98.0+%, TCI America™
Supplier: TCI America P123625G
Specifications
| Phenyl Chlorothionoformate | |
| Yellow | |
| C7H5ClOS | |
| 25 g | |
| o-phenyl carbonochloridothioate, phenyl chlorothionoformate, o-phenyl chlorothioformate, phenyl chlorothioformate, phenyl thioxochloroformate, phenyl chlorothionocarbonate, phenoxythiocarbonyl chloride, chlorothioformic acid phenyl ester, o-phenyl chlorothionoformate, o-phenyl chlorothiocarbonate | |
| ClC(=S)OC1=CC=CC=C1 | |
| 172.63 | |
| 172.63 | |
| Liquid |
| 1005-56-7 | |
| 93°C | |
| MFCD00004920 | |
| 3265 | |
| KOSYAAIZOGNATQ-UHFFFAOYSA-N | |
| phenyl chloromethanethioate | |
| 70498 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 1005-56-7 | |
| 172.63 | |
| KOSYAAIZOGNATQ-UHFFFAOYSA-N | |
| 70498 | |
| ClC(=S)OC1=CC=CC=C1 |
| C7H5ClOS | |
| MFCD00004920 | |
| o-phenyl carbonochloridothioate, phenyl chlorothionoformate, o-phenyl chlorothioformate, phenyl chlorothioformate, phenyl thioxochloroformate, phenyl chlorothionocarbonate, phenoxythiocarbonyl chloride, chlorothioformic acid phenyl ester, o-phenyl chlorothionoformate, o-phenyl chlorothiocarbonate | |
| phenyl chloromethanethioate |
Safety and Handling
TSCA : No