missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Chlorothionoformate 98.0+%, TCI America™

Catalog No. p-7135022
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12365G 5 g
P123625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12365G Supplier TCI America Supplier No. P12365G
Only null left

Chemical Identifiers

CAS 1005-56-7
Molecular Formula C7H5ClOS
Molecular Weight (g/mol) 172.63
MDL Number MFCD00004920
InChI Key KOSYAAIZOGNATQ-UHFFFAOYSA-N
Synonym o-phenyl carbonochloridothioate, phenyl chlorothionoformate, o-phenyl chlorothioformate, phenyl chlorothioformate, phenyl thioxochloroformate, phenyl chlorothionocarbonate, phenoxythiocarbonyl chloride, chlorothioformic acid phenyl ester, o-phenyl chlorothionoformate, o-phenyl chlorothiocarbonate
PubChem CID 70498
IUPAC Name phenyl chloromethanethioate
SMILES ClC(=S)OC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 93°C
Quantity 5 g
UN Number 3265
Formula Weight 172.63
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Phenyl Chlorothionoformate