missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl Acrylate (stabilized with BHT) 98.0+%, TCI America™
Supplier: TCI America P226825G
Specifications
| Phenyl Acrylate (stabilized with BHT) | |
| Yellow | |
| C9H8O2 | |
| 25 g | |
| WRAQQYDMVSCOTE-UHFFFAOYSA-N | |
| phenyl prop-2-enoate | |
| 61242 | |
| ≥98.0% (GC) |
| 937-41-7 | |
| 98°C | |
| MFCD00048145 | |
| phenyl acrylate, phenylacrylate, 2-propenoic acid, phenyl ester, phenol acrylate, phenyl-acrylate, acrylic acid phenyl ester, phenyl acrylate 5g, 2-propenoic acid,phenyl ester, acrylic acid phenyl ester, stabilized with bht | |
| C=CC(=O)OC1=CC=CC=C1 | |
| 148.161 | |
| 148.16 | |
| Liquid |
Chemical Identifiers
| 937-41-7 | |
| 148.161 | |
| WRAQQYDMVSCOTE-UHFFFAOYSA-N | |
| 61242 | |
| C=CC(=O)OC1=CC=CC=C1 |
| C9H8O2 | |
| MFCD00048145 | |
| phenyl acrylate, phenylacrylate, 2-propenoic acid, phenyl ester, phenol acrylate, phenyl-acrylate, acrylic acid phenyl ester, phenyl acrylate 5g, 2-propenoic acid,phenyl ester, acrylic acid phenyl ester, stabilized with bht | |
| phenyl prop-2-enoate |
Safety and Handling
TSCA : Yes