missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Acrylate (stabilized with BHT) 98.0+%, TCI America™

Catalog No. p-7135486
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22685G 5 g
P226825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22685G Supplier TCI America Supplier No. P22685G
Only null left

Chemical Identifiers

CAS 937-41-7
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00048145
InChI Key WRAQQYDMVSCOTE-UHFFFAOYSA-N
Synonym phenyl acrylate, phenylacrylate, 2-propenoic acid, phenyl ester, phenol acrylate, phenyl-acrylate, acrylic acid phenyl ester, phenyl acrylate 5g, 2-propenoic acid,phenyl ester, acrylic acid phenyl ester, stabilized with bht
PubChem CID 61242
IUPAC Name phenyl prop-2-enoate
SMILES C=CC(=O)OC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 98°C
Quantity 5 g
Formula Weight 148.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenyl Acrylate (stabilized with BHT)