missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl (4,6-Dimethoxy-2-pyrimidinyl)carbamate 98.0+%, TCI America™
Supplier: TCI America P218225G
Specifications
| Phenyl (4,6-Dimethoxy-2-pyrimidinyl)carbamate | |
| 120°C | |
| C13H13N3O4 | |
| 25 g | |
| (4,6-Dimethoxy-2-pyrimidinyl)carbamic Acid Phenyl Ester | |
| COC1=CC(=NC(=N1)NC(=O)OC2=CC=CC=C2)OC | |
| 275.264 | |
| 275.26 | |
| Crystalline Powder |
| 89392-03-0 | |
| White-Yellow | |
| MFCD02947731 | |
| 3077 | |
| MESPVSMSORHLAX-UHFFFAOYSA-N | |
| phenyl N-(4,6-dimethoxypyrimidin-2-yl)carbamate | |
| 10945667 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 89392-03-0 | |
| 275.264 | |
| MESPVSMSORHLAX-UHFFFAOYSA-N | |
| 10945667 | |
| COC1=CC(=NC(=N1)NC(=O)OC2=CC=CC=C2)OC |
| C13H13N3O4 | |
| MFCD02947731 | |
| (4,6-Dimethoxy-2-pyrimidinyl)carbamic Acid Phenyl Ester | |
| phenyl N-(4,6-dimethoxypyrimidin-2-yl)carbamate |
Safety and Handling
TSCA : Yes