missing translation for 'onlineSavingsMsg'
Learn More

Phenyl (4,6-Dimethoxy-2-pyrimidinyl)carbamate 98.0+%, TCI America™

Catalog No. p-7135445
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21825G 5 g
P218225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21825G Supplier TCI America Supplier No. P21825G

Chemical Identifiers

CAS 89392-03-0
Molecular Formula C13H13N3O4
Molecular Weight (g/mol) 275.264
MDL Number MFCD02947731
InChI Key MESPVSMSORHLAX-UHFFFAOYSA-N
Synonym (4,6-Dimethoxy-2-pyrimidinyl)carbamic Acid Phenyl Ester
PubChem CID 10945667
IUPAC Name phenyl N-(4,6-dimethoxypyrimidin-2-yl)carbamate
SMILES COC1=CC(=NC(=N1)NC(=O)OC2=CC=CC=C2)OC

Specifications

Melting Point 120°C
Color White-Yellow
Quantity 5 g
UN Number 3077
Formula Weight 275.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl (4,6-Dimethoxy-2-pyrimidinyl)carbamate