missing translation for 'onlineSavingsMsg'
Learn More

Phenyl 1-Hydroxy-2-naphthoate 98.0+%, TCI America™

Catalog No. H0797500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0797500G 500 g
H079725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0797500G Supplier TCI America Supplier No. H0797500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 132-54-7
Molecular Formula C17H12O3
Molecular Weight (g/mol) 264.28
MDL Number MFCD00014310
InChI Key QHDYIMWKSCJTIM-UHFFFAOYSA-N
Synonym phenyl 1-hydroxy-2-naphthoate, 1-hydroxy-2-naphthoic acid phenyl ester, 2-naphthalenecarboxylic acid, 1-hydroxy-, phenyl ester, phenyl 1-hydroxy-2-naphthalenecarboxylate, unii-0749hk9b6b, phenyl-1-hydroxy-2-naphthate, phenyl-1-hydroxy-2-naphthoate, 1-hydroxy-2-naphthalenecarboxylic acid phenyl ester, aon phenyl ester, 1-hydroxy-2-naphthoic
PubChem CID 67236
IUPAC Name phenyl 1-hydroxynaphthalene-2-carboxylate
SMILES C1=CC=C(C=C1)OC(=O)C2=C(C3=CC=CC=C3C=C2)O

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 500 g
Formula Weight 264.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl 1-Hydroxy-2-naphthoate