CAS RN 132-54-7
IUPAC Name:
phenyl 1-hydroxynaphthalene-2-carboxylate
Synonyms:
phenyl 1-hydroxynaphthalene-2-carboxylate
Molecular Weight (g/mol):
264.28
Molecular Formula:
C17H12O3
InChi Key:
QHDYIMWKSCJTIM-UHFFFAOYSA-N
SMILES:
OC1=C2C=CC=CC2=CC=C1C(=O)OC1=CC=CC=C1
1
–
1
of
1
results
1
Phenyl 1-Hydroxy-2-naphthoate 98.0+%, TCI America™
CAS: 132-54-7 Molecular Formula: C17H12O3 Molecular Weight (g/mol): 264.28 MDL Number: MFCD00014310 InChI Key: QHDYIMWKSCJTIM-UHFFFAOYSA-N Synonym: phenyl 1-hydroxy-2-naphthoate,1-hydroxy-2-naphthoic acid phenyl ester,2-naphthalenecarboxylic acid, 1-hydroxy-, phenyl ester,phenyl 1-hydroxy-2-naphthalenecarboxylate,unii-0749hk9b6b,phenyl-1-hydroxy-2-naphthate,phenyl-1-hydroxy-2-naphthoate,1-hydroxy-2-naphthalenecarboxylic acid phenyl ester,aon phenyl ester,1-hydroxy-2-naphthoic PubChem CID: 67236 IUPAC Name: phenyl 1-hydroxynaphthalene-2-carboxylate SMILES: C1=CC=C(C=C1)OC(=O)C2=C(C3=CC=CC=C3C=C2)O