missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenoxydiphenylphosphine 98.0+%, TCI America™
Supplier: TCI America P184325G
Specifications
| Phenoxydiphenylphosphine | |
| Yellow | |
| C18H15OP | |
| Phenyl Diphenylphosphinite | |
| C1=CC=C(C=C1)OP(C2=CC=CC=C2)C3=CC=CC=C3 | |
| 278.291 | |
| 278.29 | |
| Liquid |
| 13360-92-4 | |
| 170°C | |
| 25 g | |
| UPDNYUVJHQABBS-UHFFFAOYSA-N | |
| phenoxy(diphenyl)phosphane | |
| 11543680 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 13360-92-4 | |
| 278.291 | |
| Phenyl Diphenylphosphinite | |
| phenoxy(diphenyl)phosphane |
| C18H15OP | |
| UPDNYUVJHQABBS-UHFFFAOYSA-N | |
| 11543680 | |
| C1=CC=C(C=C1)OP(C2=CC=CC=C2)C3=CC=CC=C3 |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator