accessibility menu, dialog, popup

UserName

CAS RN 13360-92-4

OP

CAS RN 13360-92-4

IUPAC Name: phenyl diphenylphosphinite
Synonyms: phenyl diphenylphosphinite
Molecular Weight (g/mol): 278.29
Molecular Formula: C18H15OP
InChi Key: UPDNYUVJHQABBS-UHFFFAOYSA-N
SMILES: O(P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results