missing translation for 'onlineSavingsMsg'
Learn More

Phenoxybenzamine Hydrochloride 98.0+%, TCI America™

Catalog No. D01581G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D01581G 1 g
D015825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D01581G Supplier TCI America Supplier No. D01581G

Chemical Identifiers

CAS 63-92-3
Molecular Formula C18H23Cl2NO
Molecular Weight (g/mol) 340.288
MDL Number MFCD00055152
InChI Key VBCPVIWPDJVHAN-UHFFFAOYSA-N
Synonym phenoxybenzamine hydrochloride, dibenzyline, phenoxybenzamine hcl, n-benzyl-n-2-chloroethyl-1-phenoxypropan-2-amine hydrochloride, dibenzyline hydrochloride, phenoxybenzamine.hcl, bensylyt nen, phenoxybenzamine chloride, benzene methanamine
PubChem CID 5284441
ChEBI CHEBI:8078
IUPAC Name N-benzyl-N-(2-chloroethyl)-1-phenoxypropan-2-amine;hydrochloride
SMILES CC(COC1=CC=CC=C1)N(CCCl)CC2=CC=CC=C2.Cl

Specifications

Melting Point 139°C
Color White
Quantity 1 g
Formula Weight 340.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenoxybenzamine Hydrochloride
EINECSNumber (3)-2716
RTECSNumber DP3750000
TSCA No