missing translation for 'onlineSavingsMsg'
Learn More

Phenoxathiin 98.0+%, TCI America™

Catalog No. P12965G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12965G 5 g
P129625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12965G Supplier TCI America Supplier No. P12965G
Only null left

Chemical Identifiers

CAS 262-20-4
Molecular Formula C12H8OS
Molecular Weight (g/mol) 200.26
MDL Number MFCD00046933
InChI Key GJSGGHOYGKMUPT-UHFFFAOYSA-N
Synonym phenoxathiin, dibenzothioxin, dibenzooxathiane, phenothioxin, phenoxthin, phenoxathiane, phenoxathine, phenoxathrin, 1,4-dibenzothioxine, usaf do-17
PubChem CID 9217
IUPAC Name phenoxathiine
SMILES O1C2=CC=CC=C2SC2=CC=CC=C12

Specifications

Melting Point 57°C
Color White
Boiling Point 311°C
Quantity 5 g
Formula Weight 200.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenoxathiin
RTECSNumber SP6825000
TSCA Yes