accessibility menu, dialog, popup

UserName

OS

CAS RN 262-20-4

IUPAC Name: phenoxathiine
Synonyms: phenothioxin
Molecular Weight (g/mol): 200.26
Molecular Formula: C12H8OS
InChi Key: GJSGGHOYGKMUPT-UHFFFAOYSA-N
SMILES: O1C2=CC=CC=C2SC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results