missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein TS, Reagents

Catalog No. 01334114
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1 L
4 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334114 100 mL
01334113 1 L
01334112 250 mL
01334111 4 L
01334110 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01334114 Supplier Labchem Inc Supplier No. CP260700100A
Only null left

Phenolphthalein TS, Meets USP specifications, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 7732-18-5 , 67-56-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
IUPAC Name ethanol
SMILES CCO

Specifications

Boiling Point 77°C
Grade USP
Packaging Poly Bottle
Specific Gravity 0.7861
Color Colorless
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Phenolphthalein TS