missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenolphthalein Indicator, 1% (w/v) in Pyridine, Ricca Chemical
$175.53 - $283.57
Chemical Identifiers
| CAS | 110-86-1 |
|---|---|
| Molecular Formula | C5H5N |
| Molecular Weight (g/mol) | Mixture |
| InChI Key | JUJWROOIHBZHMG-UHFFFAOYSA-N |
| PubChem CID | 1049 |
| ChEBI | CHEBI:16227 |
| IUPAC Name | pyridine |
| SMILES | C1=CC=NC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
R5627000100
|
Ricca Chemical Company
R5627000100C |
100 mL | Amber Glass Bottle |
Each for $175.53
|
|
||||
|
5627-16
|
Ricca Chemical Company
562716 |
500 mL | Amber Glass Bottle |
Each for $232.97
|
|
||||
|
5627-32
|
Ricca Chemical Company
562732 |
1 L | Amber Glass Bottle |
Each for $283.57
|
|
||||
Chemical Identifiers
| 110-86-1 | |
| Mixture | |
| 1049 | |
| pyridine |
| C5H5N | |
| JUJWROOIHBZHMG-UHFFFAOYSA-N | |
| CHEBI:16227 | |
| C1=CC=NC=C1 |
Specifications
| 110-86-1,77-09-8 | |
| 100.0,1.5 | |
| C5H5N | |
| C1=CC=NC=C1 | |
| Mixture | |
| CHEBI:16227 | |
| Indicator | |
| Pyridine | |
| 8.5 | |
| Liquid |
| 97.0,0.5 | |
| 1% (w/v) | |
| JUJWROOIHBZHMG-UHFFFAOYSA-N | |
| pyridine | |
| 1049 | |
| 1% W/V | |
| Amber Glass Bottle | |
| Colorless to Yellow | |
| 100 mL | |
| Phenolphthalein Indicator |