missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenolphthalein Indicator, 1% (w/v) in Pyridine, Ricca Chemical
Supplier: Ricca Chemical Company 562732
Specifications
Phenolphthalein Indicator | |
97.0,0.5 | |
1% (w/v) | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
pyridine | |
1049 | |
Amber Glass Bottle | |
Colorless to Yellow | |
Liquid |
110-86-1,77-09-8 | |
100.0,1.5 | |
C5H5N | |
C1=CC=NC=C1 | |
79.102 | |
CHEBI:16227 | |
Pyridine | |
1 L |
Chemical Identifiers
110-86-1 | |
79.102 | |
1049 | |
pyridine |
C5H5N | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
CHEBI:16227 | |
C1=CC=NC=C1 |