Learn More
Fisherbrand™ Phenolic Standing Height Sink Cabinet
Fisherbrand™ Phenolic Casework is designed to rugged use in wet areas where corrosion or rust are of concern.
 View more versions of this product
Catalog No. FBPH18148P
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Rust, rot, mold resistant construction.
  • Cabinets will not absorb odors, are easy to clean and are virtually maintenance free.
  • Standard cabinet construction is from 0.5 in. thick chemically resistant phenolic panels in a full overlay style.
  • All colors have a solid black core
Specifications
Specifications
48 in.
Sink Cabinet
88.6 cm
121.9 cm
Standing Height
Platinum
2 years
22 in.
55.9 cm
34.875 in.
2
Phenolic
Product Suggestions
Product Suggestions