missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenol, ultrapure, 99%, unstab.

Catalog No. AAJ6401122
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6401122 100 g
AAJ6401136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6401122 Supplier Thermo Scientific Chemicals Supplier No. J6401122

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-95-2 | C6H6O | 94.11 g/mol

Phenol can be used for a wide variety of molecular biology applications, including nucleic acid extractions using standard phenol-chloroform protocols. Epidermal strips from the abaxial side of leaves of Commelina communis were stirred in phenol in a procedure to separate the monosaccharides from the epidermis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenol can be used for a wide variety of molecular biology applications, including nucleic acid extractions using standard phenol-chloroform protocols. Epidermal strips from the abaxial side of leaves of Commelina communis were stirred in phenol in a procedure to separate the monosaccharides from the epidermis.

Solubility
Soluble in water (82 g/L at 20°C).

Notes
Air sensitive. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Melting Point 40°C to 43°C
Color White
Density 1.071
Boiling Point 181°C
Flash Point 79°C (175°F)
Odor Sweet
Quantity 100 g
UN Number UN1671
Beilstein 969616
Sensitivity Air Sensitive
Merck Index 14,7241
Solubility Information Soluble in water (82g/L at 20°C).
Formula Weight 94.11
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Phenol, unstabilized
Grade Ultrapure
Show More Show Less
Hazard Category H301+H311+H331-H314-H335-H341-H373
Hazard Statement GHS H Statement
H301-H311-H331-H314-H341-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: PHENOL, SOLID
EINECSNumber 203-632-7
RTECSNumber SJ3325000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only