missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol, Saturated (pH 6.6/7.9, Liq.), Fisher BioReagents™
Description
Phenol, Saturated (pH 6.6/7.9, Liq.)
Phenol saturated with Tris buffer is used in DNA extraction procedures.
Specifications
Specifications
| Assay Percent Range | ≥99.0 |
| UN Number | 2821 |
| Percent Purity | ≥99.0% |
| Identification | Pass Test |
| Packaging | Coated Amber Glass/Poison Pack |
| Quantity | 100 mL |
| Physical Form | Liquid |
| Chemical Name or Material | Phenol, Saturated |
| Grade | DNase- and RNase-Free |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Combustible liquid. Causes burns by all exposure routes. Possible risks of irreversible effects. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. NFPA Health:3 Flammability:2 Instability:0 |
| DOTInformation | DOT Class 6.1, : Poison |
| Recommended Storage | 4°C |