missing translation for 'onlineSavingsMsg'
Learn More

Phenol, Saturated (pH 6.6/7.9, Liq.), Fisher BioReagents™

Catalog No. BP1750I100
Change view
Click to view available options
Quantity:
100 mL
400 mL
Packaging:
Coated Amber Glass/Poison Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP1750I100 100 mL Coated Amber Glass/Poison Pack
BP1750I400 400 mL Coated Amber Glass/Poison Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP1750I100 Supplier Fisher BioReagents Supplier No. BP1750I 100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Phenol, Saturated (pH 6.6/7.9, Liq.)

Phenol saturated with Tris buffer is used in DNA extraction procedures.

Chemical Identifiers

CAS 108-95-2 , 6381-92-6 , 77-86-1
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Assay Percent Range ≥99.0
UN Number 2821
Percent Purity ≥99.0%
Identification Pass Test
Packaging Coated Amber Glass/Poison Pack
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Phenol, Saturated
Grade DNase- and RNase-Free
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Combustible liquid. Causes burns by all exposure routes. Possible risks of irreversible effects. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:3
Flammability:2
Instability:0
DOTInformation DOT Class 6.1, : Poison
Recommended Storage 4°C