missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red, 0.04%, Aqueous, pH 6.8 to 8.4 Yellow to Red, Certified, LabChem™

Supplier:  LabChem LC182601

 View more versions of this product

Catalog No. LC182601

Only null left

Description

Description

Specifications

Specifications

Phenol Red
34487-61-1 , 7732-18-5
C19H13NaO5S
Soluble in water
HKHYOKBQJILTEI-UHFFFAOYSA-M
sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
23686673
Certified
Nitrogen oxides; Carbon monoxide; Carbon dioxide
500 mL
0.04%, Aqueous, pH 6.8 to 8.4 Yellow to Red
99.96,0.04
C19H13O5SNa
Passes Test
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]
376.358
376.36
Poly Bottle
Red
Liquid

Chemical Identifiers

34487-61-1
376.358
23686673
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]
C19H13NaO5S
HKHYOKBQJILTEI-UHFFFAOYSA-M
sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions