missing translation for 'onlineSavingsMsg'
Learn More

Phenol, Loose Crystal, Technical, Spectrum™ Chemical

Catalog No. 18612232
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
2.5 kg
25 g
45 kg
Packaging:
Amber Glass Bottle
Catalog No. Quantity Packaging
18612232 45 kg Amber Glass Bottle
18612231 25 g Amber Glass Bottle
18612230 1 kg Amber Glass Bottle
18612233 2.5 kg Amber Glass Bottle
4 options

Catalog No. 18612232

Supplier: Spectrum Chemical Mfg Cor P106945KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-95-2

Specifications

CAS Max % 1
Boiling Point 182°C
Packaging Amber Glass Bottle
Quantity 45 kg
Grade Technical
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.