missing translation for 'onlineSavingsMsg'
Learn More

Phenol, Loose Crystal, Technical, Spectrum™ Chemical

Catalog No. 18612231
Change view
Click to view available options
Quantity:
25 g
1 kg
2.5 kg
45 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612231 25 g Amber Glass Bottle
18612230 1 kg Amber Glass Bottle
18612233 2.5 kg Amber Glass Bottle
18612232 45 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18612231 Supplier Spectrum Chemical Mfg Cor Supplier No. P106925GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 108-95-2

Specifications

CAS Max % 1
Boiling Point 182°C
Packaging Amber Glass Bottle
Quantity 25 g
Grade Technical
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.